missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenyl isocyanate, 97%, Thermo Scientific™

Product Code. 10295690 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10295690 5 g Glass bottle 5g
10082100 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10295690 Supplier Thermo Scientific Acros Supplier No. 170320050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 16744-98-2
Molecular Formula C7H4FNO
Molecular Weight (g/mol) 137.113
MDL Number MFCD00001996
InChI Key VZNCSZQPNIEEMN-UHFFFAOYSA-N
Synonym 2-fluorophenyl isocyanate, o-fluorophenyl isocyanate, 2-fluorophenylisocyanate, benzene, 1-fluoro-2-isocyanato, isocyanic acid 2-fluorophenyl ester, 2-fluorobenzenisocyanate, paragos 331045, labotest-bb ltbb005297, pubchem4455, 2-flurophenylisocyanate
PubChem CID 85588
IUPAC Name 1-fluoro-2-isocyanatobenzene
SMILES C1=CC=C(C(=C1)N=C=O)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Brown
Density 1.2220g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula FC6H4NCO
Refractive Index 1.5115 to 1.5135 (20°C, 589nm)
Quantity 5 g
Formula Weight 137.11
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 2-Fluorophenyl isocyanate, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.