missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenyl isocyanate, 98%

Product Code. 11309619
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11309619 1 g 1g
11418067 5 g 5g
11428067 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11309619 Supplier Thermo Scientific Alfa Aesar Supplier No. A17466.03
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 16744-98-2 | C7H4FNO | 137.113 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16744-98-2
Molecular Formula C7H4FNO
Molecular Weight (g/mol) 137.113
MDL Number MFCD00001996
InChI Key VZNCSZQPNIEEMN-UHFFFAOYSA-N
Synonym 2-fluorophenyl isocyanate, o-fluorophenyl isocyanate, 2-fluorophenylisocyanate, benzene, 1-fluoro-2-isocyanato, isocyanic acid 2-fluorophenyl ester, 2-fluorobenzenisocyanate, paragos 331045, labotest-bb ltbb005297, pubchem4455, 2-flurophenylisocyanate
PubChem CID 85588
IUPAC Name 1-fluoro-2-isocyanatobenzene
SMILES C1=CC=C(C(=C1)N=C=O)F

Specifications

Boiling Point 56°C to 58°C (10 mmHg)
Flash Point 50°C (122°F)
Odor Pungent, Makes Eyes Water
Quantity 1 g
UN Number UN3080
Beilstein 1210127
Formula Weight 137.11
Percent Purity 98%
Chemical Name or Material 2-Fluorophenyl isocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.