missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenylacetic acid, 98%, ACROS Organics™

Product Code. 10368620 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
10g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10368620 10g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10368620 Supplier Thermo Scientific Acros Supplier No. 197520100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 451-82-1
CAS Min % 97.5
CAS Max % 100.0
Melting Point 61°C to 65°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C8H7FO2
Linear Formula FC6H4CH2CO2H
MDL Number MFCD00004315
Quantity 10 g
Beilstein 09, III, 2260
Synonym 2-fluorophenylacetic acid, 2-2-fluorophenyl acetic acid, o-fluorophenylacetic acid, benzeneacetic acid, 2-fluoro, 2-fluorophenyl acetic acid, 2-fluorophenylaceticacid, acetic acid, o-fluorophenyl, pubchem4164, 2fluorophenylacetic acid
InChI Key RPTRFSADOICSSK-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)CC(=O)O)F
IUPAC Name 2-(2-fluorophenyl)acetic acid
Molecular Weight (g/mol) 154.14
PubChem CID 67979
Formula Weight 154.14
Percent Purity ≥97.5%
Physical Form Shiny Crystals or Crystalline Powder
Chemical Name or Material 2-Fluorophenylacetic acid, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.