missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenylacetone, 98%

Product Code. 15408098
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15408098 5 g 5g
15418098 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15408098 Supplier Thermo Scientific Alfa Aesar Supplier No. H61535.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2836-82-0 | C9H9FO | 152.168 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2836-82-0
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.168
MDL Number MFCD00000325
InChI Key BANVZEUCJHUPOI-UHFFFAOYSA-N
Synonym 2-fluorophenylacetone, 1-2-fluorophenyl propan-2-one, 1-2-fluorophenyl acetone, 2-fluorophenyl acetone, o-fluorophenylacetone, 2-propanone, 1-2-fluorophenyl, acmc-20anub, ksc497e4p, 1-2-fluorophenyl acetone #
PubChem CID 76086
IUPAC Name 1-(2-fluorophenyl)propan-2-one
SMILES CC(=O)CC1=CC=CC=C1F

Specifications

Color Yellow
Density 1.077 g/mL
Boiling Point 47°C
Flash Point 85°C (185°F)
Refractive Index 1.5
Quantity 5 g
Formula Weight 152.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Fluorophenylacetone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.