missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorothiophenol, 97%

Product Code. 11491577
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491577 5 g 5g
11481577 1 g 1g
11401587 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491577 Supplier Thermo Scientific Alfa Aesar Supplier No. B24995.06
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2557-78-0 | C6H4FS | 127.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2557-78-0
Molecular Formula C6H4FS
Molecular Weight (g/mol) 127.16
MDL Number MFCD00041419
InChI Key WJTZZPVVTSDNJJ-UHFFFAOYSA-M
Synonym 2-fluorothiophenol, o-fluorothiophenol, o-fluorobenzenethiol, 2-fluorobenzene-1-thiol, fluorothiophenol, o-fluoro thiophenol, 2-fluoro-thiophenol, 2-fluoro-benzenethiol, 2-fluorobenzene thiol, pubchem5428
PubChem CID 520216
SMILES FC1=CC=CC=C1[S-]

Specifications

Density 1.203
Boiling Point 164°C
Flash Point 56°C (133°F)
Odor Stench
Refractive Index 1.555
Quantity 5 g
UN Number UN3071
Beilstein 2039768
Sensitivity Air Sensitive
IUPAC Name 2-fluorobenzenethiol
Formula Weight 128.17
Percent Purity 97%
Chemical Name or Material 2-Fluorothiophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.