missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-1,4-naphthoquinone, TRC

Product Code. 30009417
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009417 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30009417 Supplier Toronto Research Chemicals Supplier No. TRCH9495355G
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 83-72-7
Molecular Formula C10 H6 O3
Quantity 5 g
Synonym 2-Hydroxy-1,4-naphthoquinone, 1,4-Naphthalenedione, 2-hydroxy- (9CI, ACI), 1,4-Naphthoquinone, 2-hydroxy- (6CI, 8CI), 2-Hydroxy-1,4-naphthalenedione (ACI), 1,4-Dihydro-1,4-dioxo-2-hydroxynaphthalene, 2-Hydroxy-1,4-dihydronaphthalene-1,4-dione, 2-Hydroxy-1,4-naphthoquinone, 2-Hydroxynaphthoquinone, C.I. 75480, C.I. Natural Orange 6, Flower of Paradise, Henna, Henna (dye), Lawsone, Mehendi, Mendi, NSC 27285, NSC 52500, NSC 8625, Pakarli, Q 0, Quino Power LSN, Quinoexter 2014, Quinoexter QE 2014
InChI Formula InChI=1S/C10H6O3/c11-8-5-9(12)10(13)7-4-2-1-3-6(7)8/h1-5,12H
SMILES OC1=CC(=O)c2ccccc2C1=O
IUPAC Name 2-hydroxynaphthalene-1,4-dione
Molecular Weight (g/mol) 174.15
Formula Weight 174.0317
Chemical Name or Material Lawsone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.