missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-3-methoxybenzoic acid, 98+%

Product Code. p-7043625
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11391367

Brand: Thermo Scientific Alfa Aesar L01658.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 877-22-5 | C8H8O4 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877-22-5
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00002445
InChI Key AUZQQIPZESHNMG-UHFFFAOYSA-N
Synonym 3-methoxysalicylic acid, o-vanillic acid, acide orthovanillique, benzoic acid, 2-hydroxy-3-methoxy, m-anisic acid, 2-hydroxy, 3-hydroxy-m-anisic acid, unii-3j31y0e2im, 2-hydroxy-3-methoxy-benzoic acid, 3-methoxysalicyclic acid, 2-hydroxy-3-methoxy benzoic acid
PubChem CID 70140
ChEBI CHEBI:68496
IUPAC Name 2-hydroxy-3-methoxybenzoic acid
SMILES COC1=CC=CC(C(O)=O)=C1O

Specifications

Melting Point 151°C to 152°C
Quantity 5 g
Beilstein 2209642
Formula Weight 168.15
Percent Purity ≥98%
Chemical Name or Material 2-Hydroxy-3-methoxybenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.