missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-3-nitrobenzaldehyde, 98%

Product Code. p-7027134
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11460058

Brand: Thermo Scientific Alfa Aesar B23658.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5274-70-4 | C7H5NO4 | 167.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5274-70-4
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00041874
InChI Key NUGOTBXFVWXVTE-UHFFFAOYSA-N
Synonym 3-nitrosalicylaldehyde, salicylaldehyde, 3-nitro, benzaldehyde, 2-hydroxy-3-nitro, 3-nitro-2-hydroxybenzaldehyde, 3-nitrosalicaldehyde, pubchem7773, 2-hydroxy-nitrobenzaldehyde, acmc-209l1g, 3-nitrosalicylaidehyde, benzaldehyde, 2-hydroxynitro
PubChem CID 78934
IUPAC Name 2-hydroxy-3-nitrobenzaldehyde
SMILES C1=CC(=C(C(=C1)[N+](=O)[O-])O)C=O

Specifications

Melting Point 109°C to 110°C
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 167.12
Percent Purity 98%
Chemical Name or Material 2-Hydroxy-3-nitrobenzaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.