missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4,6-dimethoxybenzoic acid, 97%, Thermo Scientific™

Product Code. 11364584
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11364584

Brand: Thermo Scientific Alfa Aesar L01388.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Hydroxy-4,6-dimethoxybenzoic acid is a reagent used in the synthesis of rac Zearalenone-d6. rac Zearalenone-d6 s an estrogenic mycotoxin produced by Fusarium fungi commonly found in grains. One of a group of compounds known as resorcylic acid lactones.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3187-19-7
Molecular Formula C9H10O5
Molecular Weight (g/mol) 198.174
MDL Number MFCD00013977
InChI Key LEIJLTBRQHFMDL-UHFFFAOYSA-N
Synonym 4,6-dimethoxysalicylic acid, 4,6-dimethoxy-2-hydroxybenzoic acid, benzoic acid, 2-hydroxy-4,6-dimethoxy, acmc-1covk, 2-hydroxy-4,6-dimethoxybenzoicacid, 2,4-dimethoxy-6-hydroxybenzoic acid, 2-hydroxy-4,6-dimethoxy benzoic acid, 2-hydroxy-4,6-dimethoxy-benzoic acid, benzoic acid,2-hydroxy-4,6-dimethoxy
PubChem CID 76656
IUPAC Name 2-hydroxy-4,6-dimethoxybenzoic acid
SMILES COC1=CC(=C(C(=C1)OC)C(=O)O)O

Specifications

Melting Point 160°C to 162°C
Quantity 1 g
Beilstein 2213838
Formula Weight 198.17
Percent Purity 97%
Chemical Name or Material 2-Hydroxy-4,6-dimethoxybenzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.