missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methoxybenzoic acid, MedChemExpress

Product Code. 16431969
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16431969 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16431969 Supplier Medchem Express Supplier No. HY75625
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 2-Hydroxy-4-methoxybenzoic acid is a derivative of methoxybenzoic. 2-Hydroxy-4-methoxybenzoic is a potential biomarker.

2-Hydroxy-4-methoxybenzoic acid is a derivative of methoxybenzoic. 2-Hydroxy-4-methoxybenzoic is a potential biomarker.

Specifications

Chemical Name or Material 2-Hydroxy-4-methoxybenzoic acid
CAS 2237-36-7
Color White to Off-White
Purity Grade Notes Research
Molecular Formula C8H8O4
Quantity 1 g
Synonym 4-Methoxysalicylic Acid
Solubility Information DMSO : 50 mg/mL (297.35 mM; Need ultrasonic)
SMILES O=C(O)C1=C(O)C=C(OC)C=C1
Molecular Weight (g/mol) 168.15
Formula Weight 168.15
Percent Purity 99.55%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.