missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxy-4'-methylacetophenone, MedChemExpress

Product Code. 16487816
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487816 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16487816 Supplier Medchem Express Supplier No. HY34204

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress 2'-Hydroxy-4'-methylacetophenone, a phenolic compound isolated from Angelicae koreana roots possesses acaricidal property. It could be used in the preparation of 4’-methyl-2’-[(p-tolylsulfonyl) oxy] acetophenone.

2'-Hydroxy-4'-methylacetophenone, a phenolic compound isolated from Angelicae koreana roots possesses acaricidal property. It could be used in the preparation of 4’-methyl-2’-[(p-tolylsulfonyl) oxy] acetophenone.

Specifications

Chemical Name or Material 2'-Hydroxy-4'-methylacetophenone
CAS 6921-64-8
Color Colorless to Light Yellow
Purity Grade Notes Research
Molecular Formula C9H10O2
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (665.91 mM; Need ultrasonic)
SMILES CC(C1=CC=C(C)C=C1O)=O
Molecular Weight (g/mol) 150.17
Formula Weight 150.17
Percent Purity 96.85%
Grade Research
Physical Form Liquid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.