missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-nitrobenzoic acid, 97%

Product Code. 10613795 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10613795 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10613795 Supplier Thermo Scientific Acros Supplier No. 425870250
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 619-19-2 | C7H5NO5 | 183.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 619-19-2
CAS Min % 96.0
CAS Max % 100.0
Melting Point 235°C to 239°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C7H5NO5
Linear Formula HOC6H3(NO2)CO2H
MDL Number MFCD00071505
Quantity 25 g
Synonym 4-nitrosalicylic acid, 2-hydroxy-4-nitrobenzoic acid, benzoic acid, 2-hydroxy-4-nitro, 4-nitro-2-hydroxybenzoic acid, p-nitrosalicylic acid, salicylic acid, 4-nitro, 4-nitro-salicylsaure, 4-nitro-salicylsaure german, 2-hydroxy-4-nitro-benzoic acid, p-nitrosalicylsaure
InChI Key UKWUOTZGXIZAJC-UHFFFAOYSA-N
SMILES OC(=O)C1=CC=C(C=C1O)[N+]([O-])=O
IUPAC Name 2-hydroxy-4-nitrobenzoic acid
Molecular Weight (g/mol) 183.12
PubChem CID 69266
Formula Weight 183.12
Percent Purity 97%
Physical Form Crystalline Powder or Solid
Chemical Name or Material 2-Hydroxy-4-nitrobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydroxy-4-nitrobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.