missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-methoxybenzaldehyde, 98%

Product Code. 10700765 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10700765 50 g Glass bottle 50g
10082000 5 g Glass bottle 5g
10617142 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10700765 Supplier Thermo Scientific Acros Supplier No. 164160500
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 672-13-9 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 672-13-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00003332
InChI Key FZHSPPYCNDYIKD-UHFFFAOYSA-N
Synonym 5-methoxysalicylaldehyde, m-anisaldehyde, 6-hydroxy, benzaldehyde, 2-hydroxy-5-methoxy, salicylaldehyde, 5-methoxy, 6-hydroxy-m-anisaldehyde, 2-hydroxy-5-methoxy-benzaldehyde, 5-methoxy-2-hydroxybenzaldehyde, 2-hydroxy-5-methoxy benzaldehyde, 2-formyl-4-methoxyphenol, pubchem2649
PubChem CID 95695
IUPAC Name 2-hydroxy-5-methoxybenzaldehyde
SMILES COC1=CC(=C(C=C1)O)C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 4°C
Color Yellow
Density 1.2100g/mL
Boiling Point 103°C (2.5 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula HOC6H3(OCH3)CHO
Refractive Index 1.5770 to 1.579
Quantity 50 g
Beilstein 08, 244
Specific Gravity 1.21
Formula Weight 152.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Hydroxy-5-methoxybenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.