missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-nitrobenzaldehyde, 98%

Product Code. 11433983
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433983 25 g 25g
11423983 5 g 5g
11443983 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433983 Supplier Thermo Scientific Alfa Aesar Supplier No. A10264.14
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 97-51-8 | C7H5NO4 | 167.12 g/mol

2-Hydroxy-5-nitrobenzaldehyde is used in the synthetic preparation of coumarin (C755380), an pharmaceutic aid that is found in tonka beans, lavender oil, woodruff, sweet clover.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxy-5-nitrobenzaldehyde is used in the synthetic preparation of coumarin (C755380), an pharmaceutic aid that is found in tonka beans, lavender oil, woodruff, sweet clover.

Solubility
Soluble in water (partly).

Notes
Air & Light Sensitive,. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-51-8
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007337
InChI Key IHFRMUGEILMHNU-UHFFFAOYSA-N
Synonym 5-nitrosalicylaldehyde, benzaldehyde, 2-hydroxy-5-nitro, salicylaldehyde, 5-nitro, 5-nitrosaliclaldehyde, 2-hydroxy-5-nitro-benzaldehyde, 5-nitro salicylaldehdye, timtec-bb sbb003885, labotest-bb lt00920994, salicylaldehyde, 5-nitro-6ci,7ci,8ci, pubchem2052
PubChem CID 66808
IUPAC Name 2-hydroxy-5-nitrobenzaldehyde
SMILES OC1=CC=C(C=C1C=O)[N+]([O-])=O

Specifications

Melting Point 123°C to 127°C
Quantity 25 g
Beilstein 512565
Sensitivity Air and light sensitive
Solubility Information Soluble in water (partly).
Formula Weight 167.12
Percent Purity 98%
Chemical Name or Material 2-Hydroxy-5-nitrobenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.