missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-6-methoxybenzoic acid, MedChemExpress

Product Code. 16489386
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489386 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16489386 Supplier Medchem Express Supplier No. HYW017100
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 2-Hydroxy-6-methoxybenzoic acid can be used for the determination of acetylsalicylic acid and its major metabolite, salicylic acid, in animal plasma. 2-Hydroxy-6-methoxybenzoic acid exhibits significant analgesic effects.

2-Hydroxy-6-methoxybenzoic acid can be used for the determination of acetylsalicylic acid and its major metabolite, salicylic acid, in animal plasma. 2-Hydroxy-6-methoxybenzoic acid exhibits significant analgesic effects.

Specifications

Chemical Name or Material 2-Hydroxy-6-methoxybenzoic acid
CAS 3147-64-6
Color White to Off-White
Purity Grade Notes Research
Molecular Formula C8H8O4
Quantity 100 mg
Synonym 6-Methoxysalicylic acid
Solubility Information DMSO : 100 mg/mL (594.71 mM; Need ultrasonic)
SMILES COC1=CC=CC(O)=C1C(O)=O
Molecular Weight (g/mol) 168.15
Formula Weight 168.15
Percent Purity 97.96%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.