missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzophenone, 99%

Product Code. 11397546
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397546 5 g 5g
11307556 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397546 Supplier Thermo Scientific Alfa Aesar Supplier No. L11100.06
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 117-99-7 | C13H10O2 | 198.221 g/mol

2-Hydroxybenzophenone was used to quantify the metabolites of unmethylated extracts from the supernatants of Pseudomonas acidovorans cultures grown on 1,1-diphenylethylene. It reacts with 3-aminophenyl boronic acid to form macrocyclic boronates. 2-Hydroxybenzophenone forms copolymer 2-hydroxy, 3-allyl, 4,4?-dimethoxybenzophenone with methyl methacrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxybenzophenone was used to quantify the metabolites of unmethylated extracts from the supernatants of Pseudomonas acidovorans cultures grown on 1,1-diphenylethylene. It reacts with 3-aminophenyl boronic acid to form macrocyclic boronates. 2-Hydroxybenzophenone forms copolymer 2-hydroxy, 3-allyl, 4,4′-dimethoxybenzophenone with methyl methacrylate.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-99-7
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.221
MDL Number MFCD00002216
InChI Key HJIAMFHSAAEUKR-UHFFFAOYSA-N
Synonym 2-hydroxybenzophenone, o-benzoylphenol, 2-hydroxyphenyl phenyl methanone, o-hydroxybenzophenone, benzophenone, 2-hydroxy, methanone, 2-hydroxyphenyl phenyl, phenyl 2-hydroxyphenyl ketone, unii-lqv22w4v05, ortho-hydroxybenzophenone, 2-hydroxyphenyl phenylmethanone
PubChem CID 8348
IUPAC Name (2-hydroxyphenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CC=C2O

Specifications

Melting Point 38°C to 40°C
Boiling Point 171°C to 173°C (12 mmHg)
Quantity 5 g
Beilstein 1638174
Solubility Information Insoluble in water.
Formula Weight 198.22
Percent Purity 99%
Chemical Name or Material 2-Hydroxybenzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.