missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzophenone, 99%

Product Code. 10476970 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10476970 10 g Glass bottle 10g
10054803 1 g Glass bottle 1g
10113340 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10476970 Supplier Thermo Scientific Acros Supplier No. 146920100
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 117-99-7 | C13H10O2 | 198.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-99-7
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.22
MDL Number MFCD00002216
InChI Key HJIAMFHSAAEUKR-UHFFFAOYSA-N
Synonym 2-hydroxybenzophenone, o-benzoylphenol, 2-hydroxyphenyl phenyl methanone, o-hydroxybenzophenone, benzophenone, 2-hydroxy, methanone, 2-hydroxyphenyl phenyl, phenyl 2-hydroxyphenyl ketone, unii-lqv22w4v05, ortho-hydroxybenzophenone, 2-hydroxyphenyl phenylmethanone
PubChem CID 8348
IUPAC Name (2-hydroxyphenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CC=C2O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 37°C to 40°C
Color Yellow
Boiling Point 171°C to 173°C (12.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula HOC6H4COC6H5
Quantity 10 g
Beilstein 08, 155
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Formula Weight 198.22
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxybenzophenone, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydroxybenzophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.