missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzyl alcohol, 97%

Product Code. 15129213 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15129213 1 kg Glass bottle 1kg
10144000 25 g Glass bottle 25g
10245750 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15129213 Supplier Thermo Scientific Acros Supplier No. 173330010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 90-01-7 | C7H8O2 | 124.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-01-7
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00004617
InChI Key CQRYARSYNCAZFO-UHFFFAOYSA-N
Synonym salicyl alcohol, 2-hydroxybenzyl alcohol, saligenin, 2-hydroxymethyl phenol, 2-methylolphenol, o-hydroxybenzyl alcohol, saligenol, benzenemethanol, 2-hydroxy, diathesin, o-methylolphenol
PubChem CID 5146
ChEBI CHEBI:16464
IUPAC Name 2-(hydroxymethyl)phenol
SMILES OCC1=CC=CC=C1O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 83.0°C to 87.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula HOC6H4CH2OH
Quantity 1 kg
Beilstein 06,891
Fieser 17,305
Merck Index 15,8461
Solubility Information Solubility in water: 67g/L (22 c). Other solubilities: soluble in alcohol,chloroform,ether,propylene,glycol,benzene and fixed oils
Formula Weight 124.14
Percent Purity 97%
Physical Form Crystalline Flakes or Powder
Chemical Name or Material 2-Hydroxybenzyl alcohol, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydroxybenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.