missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzylamine, 98%

Product Code. 10416622 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10416622 1 g Glass bottle 1g
11955281 5 g Glass bottle 5g
2 options
This item is not returnable. View return policy

Product Code. 10416622

Brand: Thermo Scientific Acros 298890010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 932-30-9 | C7H9NO | 123.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 932-30-9
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.15
MDL Number MFCD00870498
InChI Key KPRZOPQOBJRYSW-UHFFFAOYSA-N
Synonym 2-aminomethyl phenol, 2-hydroxybenzylamine, phenol, 2-aminomethyl, o-hydroxybenzylamine, 2-hydroxyphenyl methylamine, unii-696r5n4nrm, salicylamine, aminomethyl phenol, o-aminomethylphenol, o-hydroxy-benzylamine
PubChem CID 70267
IUPAC Name 2-(aminomethyl)phenol
SMILES C1=CC=C(C(=C1)CN)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 127.0°C to 131.0°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula OHC6H4CH2NH2
Quantity 1 g
Formula Weight 123.15
Percent Purity 98%
Chemical Name or Material 2-Hydroxybenzylamine, 98%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.