missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxychalcone, 95%

Product Code. 11358835
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11358835 25 g 25g
11348835 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358835 Supplier Thermo Scientific Alfa Aesar Supplier No. L13937.14
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1214-47-7 | C15H12O2 | 224.259 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1214-47-7
Molecular Formula C15H12O2
Molecular Weight (g/mol) 224.259
MDL Number MFCD00016441
InChI Key AETKQQBRKSELEL-ZHACJKMWSA-N
Synonym 2'-hydroxychalcone, o-hydroxychalcone, e-2'-hydroxychalcone, 1-2-hydroxyphenyl-3-phenylprop-2-en-1-one, chalcone, 2'-hydroxy, ccris 7796, 1-2-hydroxyphenyl-3-phenyl-2-propen-1-one, unii-vy06dz94oc, acrylophenone, 2'-hydroxy-3-phenyl, 2-propen-1-one, 1-2-hydroxyphenyl-3-phenyl
PubChem CID 638276
ChEBI CHEBI:27916
IUPAC Name (E)-1-(2-hydroxyphenyl)-3-phenylprop-2-en-1-one
SMILES C1=CC=C(C=C1)C=CC(=O)C2=CC=CC=C2O

Specifications

Melting Point 89°C to 91°C
Quantity 25 g
Beilstein 976324
Formula Weight 224.26
Percent Purity ≥98%
Chemical Name or Material 2'-Hydroxychalcone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.