missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyphenethyl alcohol, 98%

Product Code. 10163064 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10163064 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10163064 Supplier Thermo Scientific Acros Supplier No. 182490050
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7768-28-7 | C8H10O2 | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7768-28-7
CAS Min % 97.5
CAS Max % 100.0
Color Brown-Yellow to Yellow
Density 1.1590g/mL
Boiling Point 168°C to 169°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C8H10O2
Linear Formula HOC6H4CH2CH2OH
Refractive Index 1.5560 to 1.5600 (20°C, 589nm)
MDL Number MFCD00002890
Quantity 5 g
Beilstein 06,906
Specific Gravity 1.159
Synonym 2-hydroxyphenethyl alcohol, 2-2-hydroxyethyl phenol, 2-hydroxyphenylethanol, o-2-hydroxyethyl phenol, benzeneethanol, 2-hydroxy, 2-2-hydroxyphenyl ethanol, o-hydroxyphenethyl alcohol, 2-o-hydroxyphenyl ethanol, 2-hydroxy-benzeneethanol, beta-o-hydroxyphenyl ethanol
InChI Key ABFCOJLLBHXNOU-UHFFFAOYSA-N
SMILES OCCC1=CC=CC=C1O
IUPAC Name 2-(2-hydroxyethyl)phenol
Molecular Weight (g/mol) 138.17
PubChem CID 82200
ChEBI CHEBI:64803
Formula Weight 138.17
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material 2-Hydroxyphenethyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydroxyphenethyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.