missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyphenoxyacetic acid, 98+%

Product Code. 11346706
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11346706

Brand: Thermo Scientific Alfa Aesar L03897.14

This item is not returnable. View return policy

CAS: 6324-11-4 | C8H8O4 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6324-11-4
MDL Number MFCD00015454

Specifications

Melting Point 125°C to 130°C
Molecular Formula C5H11ClNO2
Quantity 25 g
Beilstein 1948566
Synonym d-alanine ethyl ester hydrochloride, r-ethyl 2-aminopropanoate hydrochloride, h-d-ala-oet hcl, r-ethyl 2-amino-propionate hcl, ethyl 2r-2-aminopropanoate hydrochloride, h-ala-oblz. tos, h-d-ala-oet. hcl, 3-alanine ethyl ester hydrochloride, r-ethyl 2-amino-propionate hydrochloride
InChI Key CYHRNDXVTAWKMO-UHFFFAOYNA-N
SMILES CCOC(=O)C(C)N.Cl
IUPAC Name ethyl (2R)-2-aminopropanoate;hydrochloride
Molecular Weight (g/mol) 152.60
PubChem CID 10236019
Formula Weight 168.15
Percent Purity ≥98%
Chemical Name or Material 2-Hydroxyphenoxyacetic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.