missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyphenylacetic acid, 98%

Product Code. 11431648
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11431648 10 g 10g
11441648 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11431648 Supplier Thermo Scientific Alfa Aesar Supplier No. A13924.09
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 614-75-5 | C8H8O3 | 152.149 g/mol

2-Hydroxyphenylacetic Acid is an intermediate in the synthesis of atenolol and 3,4-dihydroxyphenylacetic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxyphenylacetic Acid is an intermediate in the synthesis of atenolol and 3,4-dihydroxyphenylacetic acid.

Solubility
Soluble in Methanol and Water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-75-5
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00004323
InChI Key CCVYRRGZDBSHFU-UHFFFAOYSA-N
Synonym 2-hydroxyphenylacetic acid, 2-2-hydroxyphenyl acetic acid, 2-hydroxyphenyl acetic acid, benzeneacetic acid, 2-hydroxy, 2-hydroxybenzeneacetic acid, o-hydroxyphenylacetic acid, acetic acid, o-hydroxyphenyl, 2-hydroxyphenylacetate, o-hydroxyphenyl acetic acid, ortho-hydroxyphenylacetic acid
PubChem CID 11970
ChEBI CHEBI:28478
IUPAC Name 2-(2-hydroxyphenyl)acetic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)O

Specifications

Melting Point 145°C to 147°C
Quantity 10 g
Beilstein 908000
Solubility Information Soluble in Methanol and Water.
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material 2-Hydroxyphenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.