missing translation for 'onlineSavingsMsg'
Learn More

2-Indanyl p-toluenesulfonate, 98+%, Thermo Scientific™

Product Code. 11383436
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11383436

Brand: Thermo Scientific Alfa Aesar L12916.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17783-69-6
Molecular Formula C16H16O3S
Molecular Weight (g/mol) 288.36
MDL Number MFCD00082747
InChI Key QUYCPYFNAZEWOT-UHFFFAOYSA-N
Synonym 2-indanyl p-toluenesulfonate, 2-indanyl p-toluenesulphonate, indan-2-yl tosylate, 2-indanyl para-toluenesulfonate, indan-2-yl 4-methylbenzenesulfonate, p-toluenesulfonic acid indan-2-yl ester
PubChem CID 7021474
IUPAC Name 2,3-dihydro-1H-inden-2-yl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OC1CC2=CC=CC=C2C1

Specifications

Melting Point 118°C to 121°C
Quantity 5 g
Beilstein 2147482
Sensitivity Moisture sensitive
Formula Weight 288.37
Percent Purity ≥98%
Chemical Name or Material 2-Indanyl p-toluenesulfonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.