missing translation for 'onlineSavingsMsg'
Learn More

2-Indanylboronic acid diethanolamine ester, 98%, Thermo Scientific™

Product Code. 15470857
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470857 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15470857 Supplier Thermo Scientific Alfa Aesar Supplier No. L19574.MC
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 501014-44-4
Melting Point 213°C to 219°C
Molecular Formula C13H18BNO2
MDL Number MFCD03788732
Quantity 100 mg
Synonym 2-indanylboronic acid diethanolamine ester, 2-2,3-dihydro-1h-inden-2-yl-1,3,6,2-dioxazaborocane, 2-indanylboronic acid diethanolamine cyclic ester, 2-indan-2-yl-1,3-dioxa-6-aza-2-boracyclooctane
InChI Key PLSXCXOMNZXQRU-UHFFFAOYSA-N
SMILES B1(OCCNCCO1)C2CC3=CC=CC=C3C2
IUPAC Name 2-(2,3-dihydro-1H-inden-2-yl)-1,3,6,2-dioxazaborocane
Molecular Weight (g/mol) 231.102
PubChem CID 22572023
Formula Weight 231.1
Percent Purity 98%
Chemical Name or Material 2-Indanylboronic acid diethanolamine ester
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.