missing translation for 'onlineSavingsMsg'
Learn More

2-Indanylboronic acid pinacol ester, 95%

Product Code. 11348916
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11348916 250 mg 250mg
11338916 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11348916 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' L19535.MD
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 608534-44-7 | C15H21BO2 | 244.14 g/mol

Boronic acids react with alcohols, with loss of water, to form boronic esters. With simple alcohols, the products are very susceptible to hydrolysis, but with 1,2- and 1,3-diols, the resulting cyclic boronates (1,3,2- dioxaborolanes and 1,3,2-dioxaborinanes) are stable enough to be isolated.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Boronic acids react with alcohols, with loss of water, to form boronic esters. With simple alcohols, the products are very susceptible to hydrolysis, but with 1,2- and 1,3-diols, the resulting cyclic boronates (1,3,2- dioxaborolanes and 1,3,2-dioxaborinanes) are stable enough to be isolated.

Solubility
Difficult to mix in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608534-44-7
Molecular Formula C15H21BO2
Molecular Weight (g/mol) 244.14
MDL Number MFCD03788726
InChI Key YRTKSYXHFWUPAZ-UHFFFAOYSA-N
Synonym 2-indanylboronic acid pinacol ester, 2-2,3-dihydro-1h-inden-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-indanylboronicacidpinacolester, 2-indan-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 24208876
IUPAC Name 2-(2,3-dihydro-1H-inden-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1CC2=CC=CC=C2C1

Specifications

Refractive Index 1.504
Quantity 250 mg
Solubility Information Difficult to mix in water.
Formula Weight 244.14
Percent Purity 95%
Chemical Name or Material 2-Indanylboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.