missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-4-(trifluoromethyl)aniline, 99%

Product Code. 11425337
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11425337 250 g 250g
11405337 10 g 10g
11415337 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11425337 Supplier Thermo Scientific Alfa Aesar Supplier No. B22691.30
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 163444-17-5 | C7H5F3IN | 287.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 163444-17-5
Molecular Formula C7H5F3IN
Molecular Weight (g/mol) 287.02
MDL Number MFCD02683097
InChI Key UKKWTZPXYIYONW-UHFFFAOYSA-N
Synonym 4-amino-3-iodobenzotrifluoride, 2-iodo-4-trifluoromethyl aniline, 4-trifluoromethyl-2-iodobenzenamine, benzenamine, 2-iodo-4-trifluoromethyl, 2-iodo-4-trifluoromethyl benzenamine, pubchem1632, acmc-209dpp, intermediates-zcf02228, ksc910s3b, 3-iodo-4-aminobenzotrifluoride
PubChem CID 2783322
IUPAC Name 2-iodo-4-(trifluoromethyl)aniline
SMILES NC1=C(I)C=C(C=C1)C(F)(F)F

Specifications

Melting Point 49°C to 52°C
Quantity 250 g
Sensitivity Light sensitive
Formula Weight 287.02
Percent Purity 99%
Chemical Name or Material 2-Iodo-4-(trifluoromethyl)aniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.