missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-5-methylbenzoic acid methyl ester, Thermo Scientific™

Product Code. 10179589
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10179589 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10179589 Supplier Thermo Scientific Maybridge Supplier No. MAY00280DE
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 103440-52-4
Packaging Amber Glass Bottle
Molecular Formula C9H9IO2
MDL Number MFCD12198129
Quantity 5 g
Synonym 2-iodo-5-methylbenzoic acid methyl ester, benzoic acid, 2-iodo-5-methyl-, methyl ester, benzoic acid,2-iodo-5-methyl-, methyl ester, acmc-20dzjp, methyl 6-iodo-m-toluate, methyl2-iodo-5-methylbenzoate, 4-iodo-3-methoxycarbonyltoluene, 6-iodo-m-toluic acid methyl ester, methyl 2-iodanyl-5-methyl-benzoate, methyl 2-iodo-5-methylbenzoate
InChI Key DHYMLHOXGMUIDZ-UHFFFAOYSA-N
SMILES COC(=O)C1=C(I)C=CC(C)=C1
IUPAC Name methyl 2-iodo-5-methylbenzoate
Molecular Weight (g/mol) 276.07
PubChem CID 14263946
Formula Weight 276.07
Percent Purity 95%
Chemical Name or Material 2-Iodo-5-methylbenzoic acid methyl ester
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.