missing translation for 'onlineSavingsMsg'
Learn More

2-Iodoaniline, 98%

Product Code. 10306760 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10306760 25 g Glass bottle 25g
10538660 5 g Glass bottle 5g
10493055 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10306760 Supplier Thermo Scientific Acros Supplier No. 122300250
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 615-43-0 | C6H6IN | 219.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-43-0
Molecular Formula C6H6IN
Molecular Weight (g/mol) 219.03
MDL Number MFCD00007680
InChI Key UBPDKIDWEADHPP-UHFFFAOYSA-N
Synonym o-iodoaniline, benzenamine, 2-iodo, 2-iodo aniline, 2-iodobenzenamine, o-aminoiodobenzene, aniline, o-iodo, aniline, 2-iodo, benzenamine, iodo, 2-iodo-phenylamine, 2-iodophenylamine
PubChem CID 11995
IUPAC Name 2-iodoaniline
SMILES NC1=CC=CC=C1I

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 53°C to 58°C
Color Brown to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 12, 669
Solubility Information Solubility in water: slightly soluble
Formula Weight 219.02
Percent Purity 98%
Physical Form Crystalline Needles
Chemical Name or Material 2-Iodoaniline
Show More Show Less
compliance-icons
Product Identifier
  • 2-Iodoaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.