missing translation for 'onlineSavingsMsg'
Learn More

2-Iodoanisole, 97%

Product Code. 10640463 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10640463 5 g Glass bottle 5g
10637592 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10640463 Supplier Thermo Scientific Acros Supplier No. 249680050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 529-28-2 | C7H7IO | 234.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 529-28-2
Molecular Formula C7H7IO
Molecular Weight (g/mol) 234.04
MDL Number MFCD00001039
InChI Key DVQWNQBEUKXONL-UHFFFAOYSA-N
Synonym 2-iodoanisole, o-iodoanisole, benzene, 1-iodo-2-methoxy, iodoanisole, 2-methoxyiodobenzene, o-anisyl iodide, 2-iodophenol methyl ether, anisole, o-iodo, 2-iodophenyl methyl ether, 1-iodo-2-methoxy-benzene
PubChem CID 68257
ChEBI CHEBI:16355
IUPAC Name 1-iodo-2-methoxybenzene
SMILES COC1=CC=CC=C1I

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.7900g/mL
Boiling Point 238°C to 240°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula IC6H4OCH3
Refractive Index 1.6210 to 1.623
Quantity 5 g
Merck Index 15, 5071
Specific Gravity 1.79
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform and ether
Formula Weight 234.04
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Iodoanisole, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Iodoanisole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.