missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobenzoic acid, 98+%

Product Code. 11483204
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483204 25 g 25g
11493204 100 g 100g
11403214 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11483204 Supplier Thermo Scientific Alfa Aesar Supplier No. A10563.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 88-67-5 | C7H5IO2 | 248.019 g/mol

Reagent for detection of sulfhydryl groups in proteins2-Iodobenzoic acid acts as a precursor in the preparation of 2-Iodoxybenzoic acid (IBX) and Dess-Martin periodinane, which finds application as an oxidizing agent in synthetic chemistry. It is also used in Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for detection of sulfhydryl groups in proteins2-Iodobenzoic acid acts as a precursor in the preparation of 2-Iodoxybenzoic acid (IBX) and Dess-Martin periodinane, which finds application as an oxidizing agent in synthetic chemistry. It is also used in Suzuki reaction.

Solubility
Soluble in dimethyl sulfoxide and methanol

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-67-5
Molecular Formula C7H5IO2
Molecular Weight (g/mol) 248.019
MDL Number MFCD00002419
InChI Key CJNZAXGUTKBIHP-UHFFFAOYSA-N
Synonym o-iodobenzoic acid, benzoic acid, 2-iodo, iodobenzoic acid, benzoic acid, o-iodo, usaf ek-572, 2-iodo benzoic acid, 2-iodo-benzoic acid, benzoic acid, iodo, kyselina o-jodbenzoova, o-jod-benzoesaeure
PubChem CID 6941
ChEBI CHEBI:287979
IUPAC Name 2-iodobenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)I

Specifications

Melting Point 160°C to 164°C
Density 2.25
Quantity 25 g
Beilstein 1861406
Sensitivity Light sensitive
Merck Index 14,5030
Solubility Information Soluble in dimethyl sulfoxide and methanol
Formula Weight 248.02
Percent Purity ≥98%
Chemical Name or Material 2-Iodobenzoic acid
compliance-icons compliance-icons
Product Identifier
  • 2-Iodobenzoic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Iodobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.