missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobenzoic acid, 98+%

Product Code. 11493204
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Pakningsstørrelse
11493204 100 g 100g
11483204 25 g 25g
11403214 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11493204 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. A10563.22
 Restricted Item
Estimated Shipment: 12-11-2026
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

CAS: 88-67-5 | C7H5IO2 | 248.019 g/mol

Reagent for detection of sulfhydryl groups in proteins2-Iodobenzoic acid acts as a precursor in the preparation of 2-Iodoxybenzoic acid (IBX) and Dess-Martin periodinane, which finds application as an oxidizing agent in synthetic chemistry. It is also used in Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for detection of sulfhydryl groups in proteins2-Iodobenzoic acid acts as a precursor in the preparation of 2-Iodoxybenzoic acid (IBX) and Dess-Martin periodinane, which finds application as an oxidizing agent in synthetic chemistry. It is also used in Suzuki reaction.

Solubility
Soluble in dimethyl sulfoxide and methanol

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 88-67-5
Molecular Formula C7H5IO2
Molecular Weight (g/mol) 248.019
MDL Number MFCD00002419
InChI Key CJNZAXGUTKBIHP-UHFFFAOYSA-N
Synonym o-iodobenzoic acid, benzoic acid, 2-iodo, iodobenzoic acid, benzoic acid, o-iodo, usaf ek-572, 2-iodo benzoic acid, 2-iodo-benzoic acid, benzoic acid, iodo, kyselina o-jodbenzoova, o-jod-benzoesaeure
PubChem CID 6941
ChEBI CHEBI:287979
IUPAC Name 2-iodobenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)I

Tekniske data

Melting Point 160°C to 164°C
Density 2.25
Quantity 100 g
Beilstein 1861406
Sensitivity Light sensitive
Merck Index 14,5030
Solubility Information Soluble in dimethyl sulfoxide and methanol
Formula Weight 248.02
Percent Purity ≥98%
Chemical Name or Material 2-Iodobenzoic acid

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.