missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobenzoic acid, 98%

Product Code. 10456930 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10456930 25 g Plastic bottle 25g
10657082 100 g Plastic bottle 100g
11692739 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10456930 Supplier Thermo Scientific Acros Supplier No. 122340250
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 88-67-5 | C7H5IO2 | 248.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-67-5
Molecular Formula C7H5IO2
Molecular Weight (g/mol) 248.01
InChI Key CJNZAXGUTKBIHP-UHFFFAOYSA-N
Synonym o-iodobenzoic acid, benzoic acid, 2-iodo, iodobenzoic acid, benzoic acid, o-iodo, usaf ek-572, 2-iodo benzoic acid, 2-iodo-benzoic acid, benzoic acid, iodo, kyselina o-jodbenzoova, o-jod-benzoesaeure
PubChem CID 6941
ChEBI CHEBI:287979
IUPAC Name 2-iodobenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)I

Specifications

Melting Point 161.0°C to 163.0°C
Color Brown-Orange to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula IC6H4CO2H
Quantity 25 g
Beilstein 09,363
Fieser 01,506
Merck Index 15,5074
Solubility Information Solubility in water: sparingly soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 248.01
Percent Purity 98%
Physical Form Crystals, Powder or Flakes
Chemical Name or Material 2-Iodobenzoic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Iodobenzoic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.