missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobiphenyl, 98%

Product Code. 11397076
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397076 1 g 1g
11307086 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397076 Supplier Thermo Scientific Alfa Aesar Supplier No. L06990.03
 Restricted Item
Estimated Shipment: 20-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2113-51-1 | C12H9I | 280.108 g/mol

2-Iodobiphenyl is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodobiphenyl is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2113-51-1
Molecular Formula C12H9I
Molecular Weight (g/mol) 280.108
MDL Number MFCD00039396
InChI Key QFUYDAGNUJWBSM-UHFFFAOYSA-N
Synonym 2-iodobiphenyl, 2-iodo-1,1'-biphenyl, o-iodobiphenyl, 1,1'-biphenyl, 2-iodo, o-phenyliodobenzene, 2-iodo-biphenyl, biphenyl, 2-iodo, 2-iodo-1-phenylbenzene, 1-iodo-2-phenyl-benzene, 1,1'-biphenyl, iodo
PubChem CID 75025
IUPAC Name 1-iodo-2-phenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=CC=C2I

Specifications

Density 1.59
Boiling Point 300°C
Flash Point >110°C (230°F)
Refractive Index 1.661
Quantity 1 g
Beilstein 2206820
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 280.11
Percent Purity 98%
Chemical Name or Material 2-Iodobiphenyl
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.