missing translation for 'onlineSavingsMsg'
Learn More

2-Iodotoluene, 98%

Product Code. 10568840 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10568840 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10568840 Supplier Thermo Scientific Acros Supplier No. 122430250
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 615-37-2 | C7H7I | 218.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 615-37-2
CAS Min % 97.5
CAS Max % 100.0
Color Orange to Yellow
Density 1.7100g/mL
Boiling Point 211°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H7I
Linear Formula CH3C6H4I
Refractive Index 1.6080 to 1.61
MDL Number MFCD00001042
Quantity 25 mL
Beilstein 05, 310
Specific Gravity 1.71
Synonym 2-iodotoluene, o-iodotoluene, o-tolyl iodide, benzene, 1-iodo-2-methyl, o-methyliodobenzene, toluene, o-iodo, iodotoluene, ortho-iodotoluene, 2-methyliodobenzene, unii-8ok4h85t07
InChI Key RINOYHWVBUKAQE-UHFFFAOYSA-N
SMILES CC1=CC=CC=C1I
IUPAC Name 1-iodo-2-methylbenzene
Molecular Weight (g/mol) 218.04
PubChem CID 5128
ChEBI CHEBI:39813
Formula Weight 218.04
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Iodotoluene, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.