missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylaniline, 97%

Product Code. 11467646
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11467646 500 g 500g
11447646 25 g 25g
11457646 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467646 Supplier Thermo Scientific Alfa Aesar Supplier No. A14671.36
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 643-28-7 | C9H13N | 135.21 g/mol

2-Isopropylaniline is used in the synthesis of a colorless amine-coordinated zinc complex. The reactions of 1,1?-bis(hydroxymethyl)ferrocene with 2-isopropylaniline, in the presence of a catalyst [RuCl2(PPh3)3], was studied

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Isopropylaniline is used in the synthesis of a colorless amine-coordinated zinc complex. The reactions of 1,1′-bis(hydroxymethyl)ferrocene with 2-isopropylaniline, in the presence of a catalyst [RuCl2(PPh3)3], was studied

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents, strong acids, acid chlorides, acid anhydrides. Light Sensitive
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 643-28-7
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00007720
InChI Key YKOLZVXSPGIIBJ-UHFFFAOYSA-N
Synonym 2-isopropylaniline, o-isopropylaniline, o-cumidine, 2-propan-2-yl aniline, benzenamine, 2-1-methylethyl, o-aminoisopropylbenzene, 2-isopropyl aniline, 2-aminoisopropylbenzene, aniline, o-isopropyl, o-isopropyl aniline
PubChem CID 12561
SMILES CC(C)C1=CC=CC=C1N

Specifications

Density 0.966
Boiling Point 214°C to 216°C
Flash Point 95°C (203°F)
Refractive Index 1.5485
Quantity 500 g
Beilstein 636283
Sensitivity Light sensitive
Solubility Information Insoluble in water.
IUPAC Name 2-propan-2-ylaniline
Formula Weight 135.21
Percent Purity 97%
Chemical Name or Material 2-Isopropylaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.