missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylaniline, 97%, Thermo Scientific™

Product Code. 10120552 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10120552 5 mL Glass bottle 5mL
10333812 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10120552 Supplier Thermo Scientific Acros Supplier No. 259390050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 643-28-7
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00007720
InChI Key YKOLZVXSPGIIBJ-UHFFFAOYSA-N
Synonym 2-isopropylaniline, o-isopropylaniline, o-cumidine, 2-propan-2-yl aniline, benzenamine, 2-1-methylethyl, o-aminoisopropylbenzene, 2-isopropyl aniline, 2-aminoisopropylbenzene, aniline, o-isopropyl, o-isopropyl aniline
PubChem CID 12561
SMILES CC(C)C1=CC=CC=C1N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Brown or Orange to Yellow
Density 0.9550g/mL
Boiling Point 112.0°C to 113.0°C (18.0 mmHg)
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHC6H4NH2
Refractive Index 1.5470 to 1.5500 (20°C, 589nm)
Quantity 5 mL
Specific Gravity 0.955
Solubility Information Solubility in water: insoluble
IUPAC Name 2-propan-2-ylaniline
Formula Weight 135.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Isopropylaniline
Show More Show Less
compliance-icons
Product Identifier
  • 2-Isopropylaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.