missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylphenol, 97%

Product Code. 10533131 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
Packaging:
Glass bottle
Unit Size:
500g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10533131 5 g Glass bottle 5g
10496800 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10533131 Supplier Thermo Scientific Acros Supplier No. 156680050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 88-69-7 | C9H12O | 136.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-69-7
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00002224
InChI Key CRBJBYGJVIBWIY-UHFFFAOYSA-N
Synonym 2-isopropylphenol, o-hydroxycumene, o-isopropylphenol, o-cumenol, 1-hydroxy-2-isopropylbenzene, 2-propan-2-yl phenol, 2-1-methylethyl phenol, phenol, o-isopropyl, phenol, 2-1-methylethyl, isopropylphenol
PubChem CID 6943
ChEBI CHEBI:38506
IUPAC Name 2-propan-2-ylphenol
SMILES CC(C)C1=CC=CC=C1O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 15.0°C to 16.0°C
Color Yellow
Density 1.0100g/mL
Boiling Point 212.0°C to 213.0°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (CH3)2CHC6H4OH
Refractive Index 1.5255 to 1.5275
Quantity 5 g
Beilstein 06, 504
Specific Gravity 1.01
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in 10% sodium hydroxide,soluble in isopentane,toluene,ethyl alcohol
Formula Weight 136.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Isopropylphenol, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Isopropylphenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.