missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylphenyl isocyanate, 95%, Thermo Scientific™

Product Code. 11915701 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11915701 5 g 5g
11905701 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11915701 Supplier Thermo Scientific Acros Supplier No. 441570050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 56309-56-9
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.21
InChI Key MWBSQVPLYJZPTG-UHFFFAOYSA-N
Synonym 2-isopropylphenyl isocyanate, 1-isocyanato-2-isopropylbenzene, o-isopropylphenyl isocyanate, 2-isopropylphenylisocyanate, 1-isocyanato-2-propan-2-yl benzene, 1-isocyanato-2-isopropyl-benzene, 2-methylethyl benzenisocyanate, acmc-1avxp, isopropylphenyl isocyanate
PubChem CID 91797
IUPAC Name 1-isocyanato-2-propan-2-ylbenzene
SMILES CC(C)C1=CC=CC=C1N=C=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Density 1.0190g/mL
Boiling Point 66.0°C to 67.0°C (5.0 mmHg)
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Quantity 5 g
Specific Gravity 1.019
Formula Weight 161.21
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Isopropylphenyl isocyanate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Isopropylphenyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.