missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylphenyl isocyanate, 97%

Product Code. 11392896
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392896 1 g 1g
11302906 5 g 5g
11312906 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11392896 Supplier Thermo Scientific Alfa Aesar Supplier No. L10618.03
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56309-56-9 | C10H11NO | 161.204 g/mol

2-Isopropylphenyl isocyanate is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Isopropylphenyl isocyanate is used as intermediates.

Solubility
Insoluble in water.

Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from moisture, alocohol, amines, water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56309-56-9
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.204
MDL Number MFCD00013850
InChI Key MWBSQVPLYJZPTG-UHFFFAOYSA-N
Synonym 2-isopropylphenyl isocyanate, 1-isocyanato-2-isopropylbenzene, o-isopropylphenyl isocyanate, 2-isopropylphenylisocyanate, 1-isocyanato-2-propan-2-yl benzene, 1-isocyanato-2-isopropyl-benzene, 2-methylethyl benzenisocyanate, acmc-1avxp, isopropylphenyl isocyanate
PubChem CID 91797
IUPAC Name 1-isocyanato-2-propan-2-ylbenzene
SMILES CC(C)C1=CC=CC=C1N=C=O

Specifications

Boiling Point 66°C to 67°C (5 mmHg)
Flash Point 87°C (188°F)
Quantity 1 g
UN Number UN2206
Beilstein 3588928
Formula Weight 161.21
Percent Purity 97%
Chemical Name or Material 2-Isopropylphenyl isocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.