missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylphenyl isothiocyanate, 96%

Product Code. 11494027
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494027 25 g 25g
11309599 1 g 1g
11484027 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494027 Supplier Thermo Scientific Alfa Aesar Supplier No. A19448.14
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 36176-31-5 | C10H11NS | 177.265 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36176-31-5
Molecular Formula C10H11NS
Molecular Weight (g/mol) 177.265
MDL Number MFCD00022053
InChI Key QQOOMDSEMQHMIR-UHFFFAOYSA-N
Synonym 2-isopropylphenyl isothiocyanate, 1-isopropyl-2-isothiocyanatobenzene, 1-isothiocyanato-2-propan-2-yl benzene, benzene, 1-isothiocyanato-2-1-methylethyl, 2-methylethyl benzenisothiocyanate, 2-isopropylphenylisothiocyanate, acmc-20ansf, 2-isopropylphenyl-isothiocyanate, 2-iso-propylphenyl isothiocyanate
PubChem CID 142052
IUPAC Name 1-isothiocyanato-2-propan-2-ylbenzene
SMILES CC(C)C1=CC=CC=C1N=C=S

Specifications

Density 1.05
Boiling Point 131°C to 132°C (10 mmHg)
Refractive Index 1.605
Quantity 25 g
UN Number UN2922
Beilstein 2717112
Sensitivity Moisture sensitive
Formula Weight 177.27
Percent Purity 96%
Chemical Name or Material 2-Isopropylphenyl isothiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.