missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-4,5-diphenyloxazole, 98%, Thermo Scientific™

Product Code. 11420631
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420631 25 g 25g
11410631 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420631 Supplier Thermo Scientific Alfa Aesar Supplier No. B20111.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6670-13-9
Molecular Formula C15H11NOS
Molecular Weight (g/mol) 253.319
MDL Number MFCD00173660
InChI Key NYQSTRUSYDZNHC-UHFFFAOYSA-N
Synonym 4,5-diphenyl-2-oxazolethiol, 4,5-diphenyl-1,3-oxazole-2-thiol, diphenyl-1,3-oxazole-2-thiol, 4,5-diphenyl-2-mercaptooxazole, 4,5-diphenyl-1,3-oxazole-2 3h-thione, 4,5-diphenyloxazole-2 3h-thione, 4,5-diphenyl-4-oxazoline-2-thione, 4,5-diphenyl-2,3-dihydro-1,3-oxazole-2-thione, 4,5-diphenyloxazole-2-thiol, 4,5-diphenyl-3h-oxazole-2-thione
PubChem CID 853676
IUPAC Name 4,5-diphenyl-3H-1,3-oxazole-2-thione
SMILES C1=CC=C(C=C1)C2=C(OC(=S)N2)C3=CC=CC=C3

Specifications

Melting Point 251°C to 256°C
Quantity 25 g
Sensitivity Air Sensitive
Formula Weight 253.33
Percent Purity 98%
Chemical Name or Material 2-Mercapto-4,5-diphenyloxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.