missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptophenol, 98+%

Product Code. 10520184 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10520184 5 g Glass bottle 5g
10512004 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10520184 Supplier Thermo Scientific Acros Supplier No. 385600050
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1121-24-0 | C6H6OS | 126.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1121-24-0
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.18
MDL Number MFCD00040447
InChI Key VMKYTRPNOVFCGZ-UHFFFAOYSA-N
Synonym 2-hydroxythiophenol, 2-mercaptophenol, o-mercaptophenol, phenol, 2-mercapto, 2-hydroxy thiophenol, 2-hydroxybenzenethiol, o-hydroxythiophenol, o-hydroxybenzenothiol, 2-hydroxybenzenothiol, mercaptophenol
PubChem CID 70718
IUPAC Name 2-sulfanylphenol
SMILES C1=CC=C(C(=C1)O)S

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 20.0°C
Color Green to Yellow
Density 1.2200g/mL
Boiling Point 40.0°C (300.0 mmHg)
Flash Point 97°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula SHC6H4OH
Refractive Index 1.6100 to 1.6120
Quantity 5 g
Specific Gravity 1.22
Solubility Information Solubility in water: slightly soluble.
Formula Weight 126.18
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2-Mercaptophenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Mercaptophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.