missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-1-naphthaldehyde, 99%

Product Code. 11414477
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414477 5 g 5g
11424477 25 g 25g
11434477 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414477 Supplier Thermo Scientific Alfa Aesar Supplier No. B20483.06
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. Consulta la politica di reso

CAS: 5392-12-1 | C12H10O2 | 186.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 5392-12-1
MDL Number MFCD00004004

Specifica

Melting Point 81°C to 85°C
Boiling Point 204°C to 206°C (18 mmHg)
Molecular Formula C14H10O2
Quantity 5 g
Beilstein 1868308
Sensitivity Air Sensitive
Synonym 3-phenylphthalide, 3-phenyl phthalide, phthalide, 3-phenyl, 3-phenyl-2-benzofuran-1 3h-one, 3-phenyl-1 3h-isobenzofuranone, 3-phenyl-1,3-dihydro-2-benzofuran-1-one, 1 3h-isobenzofuranone, 3-phenyl, 3-phenyl-3-hydroisobenzofuran-1-one, phenylphthalid, acmc-1ak9n
InChI Key SQFMIHCARVMICF-UHFFFAOYNA-N
SMILES C1=CC=C(C=C1)C2C3=CC=CC=C3C(=O)O2
IUPAC Name 3-phenyl-3H-2-benzofuran-1-one
Molecular Weight (g/mol) 210.23
PubChem CID 95365
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material 2-Methoxy-1-naphthaldehyde
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.