missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-methylphenol, 99%

Product Code. p-157447 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10627381

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 93-51-6 | C8H10O2 | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-51-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00002378
InChI Key PETRWTHZSKVLRE-UHFFFAOYSA-N
Synonym creosol, 4-methylguaiacol, 2-methoxy-p-cresol, homoguaiacol, p-methylguaiacol, phenol, 2-methoxy-4-methyl, p-creosol, 4-methyl guaiacol, 2-methoxy-4-cresol, 4-hydroxy-3-methoxytoluene
PubChem CID 7144
IUPAC Name 2-methoxy-4-methylphenol
SMILES CC1=CC(=C(C=C1)O)OC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 2.0°C
Color Yellow
Density 1.0900g/mL
Boiling Point 221.0°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5360 to 1.5380
Quantity 25 g
Merck Index 15, 2559
Specific Gravity 1.09
Solubility Information Solubility in water: insoluble
Formula Weight 138.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Methoxy-4-methylphenol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methoxy-4-methylphenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.