missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-nitroaniline, 98%

Product Code. 10011830 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy

Product Code. 10011830

Brand: Thermo Scientific Acros 150641000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 97-52-9 | C7H8N2O3 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-52-9
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.15
MDL Number MFCD00007363
InChI Key GVBHRNIWBGTNQA-UHFFFAOYSA-N
Synonym fast red b base, benzenamine, 2-methoxy-4-nitro, 4-nitro-o-anisidine, fast red base b, fast red b, red b base, diazo fast red b, 2-amino-5-nitroanisole, azoamine pink o, fast red 5na base
PubChem CID 7337
IUPAC Name 2-methoxy-4-nitroaniline
SMILES COC1=CC(=CC=C1N)[N+]([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 139.0°C to 142.0°C
Color Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula CH3OC6H3(NO2)NH2
Quantity 100 g
Beilstein 13, 390
Solubility Information Solubility in water: slightly soluble
Formula Weight 168.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxy-4-nitroaniline
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methoxy-4-nitroaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.