missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-nitroaniline, TRC

Product Code. 17941715
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17941715 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17941715 Supplier Toronto Research Chemicals Supplier No. TRCM2647585G
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 97-52-9
Molecular Formula C7 H8 N2 O3
Quantity 5 g
Synonym 2-Methoxy-4-nitroaniline, Benzenamine, 2-methoxy-4-nitro-, Fast Red B Base (6CI), o-Anisidine, 4-nitro- (8CI), 2-Methoxy-4-nitrobenzenamine, 1-Amino-2-methoxy-4-nitrobenzene, 2-Amino-1-methoxy-5-nitrobenzene, 2-Amino-5-nitroanisole, 2-Methoxy-4-nitroaniline, 3-Methoxy-4-aminonitrobenzene, 3-Nitro-6-aminoanisole, 4-Amino-3-methoxynitrobenzene, 4-Nitro-2-methoxyaniline, 4-Nitro-6-methoxyaniline, 4-Nitro-o-anisidine, Amarthol Fast Red B Base, Azoamine Pink O, Azoene Fast Red B Base, Brentamine Fast Red B Base, C.I. 37125, Dainichi Fast Red B Base, Daito Red Base B, Devol Red E, Diabase Red B, Fast Red 5NA Base, Fast Red B-T Base, Fast Red Base B, Hiltonil Fast Red B Base, Kako Red B Base, Kayaku Red B Base, Mitsui Red B Base, NSC 4130, Naphthanil Red B Base, Naphtoelan Red B Base, PNOA, Red B Base, Red Base Ciba V, Red Base Irga V, Red Base NB, Sanyo Fast Red B Base, Shinnippon Fast Red B Base, Showa Fast Red B Base, Symulon Red B Base, o-Methoxy-p-nitroaniline, p-Nitro-o-anisidine, 2-Methoxy-4-nitrobenzenamine, 1-Amino-2-methoxy-4-nitrobenzene, 2-Amino-1-methoxy-5-nitrobenzene, 2-Amino-5-nitroanisole, 2-Methoxy-4-nitroaniline, 3-Methoxy-4-aminonitrobenzene, 3-Nitro-6-aminoanisole, 4-Amino-3-methoxynitrobenzene, 4-Nitro-2-methoxyaniline, 4-Nitro-6-methoxyaniline, 4-Nitro-o-anisidine
InChI Formula InChI=1S/C7H8N2O3/c1-12-7-4-5(9(10)11)2-3-6(7)8/h2-4H,8H2,1H3
SMILES COc1cc(ccc1N)[N+](=O)[O-]
IUPAC Name 2-methoxy-4-nitroaniline
Molecular Weight (g/mol) 168.15
Formula Weight 168.0535
Chemical Name or Material 2-Methoxy-4-nitroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.