missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-nitrobenzenesulfonyl chloride, 97%

Product Code. 11329935
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329935 1 g 1g
11339935 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329935 Supplier Thermo Scientific Alfa Aesar Supplier No. L19902.03
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 21320-91-2 | C7H6ClNO5S | 251.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21320-91-2
Molecular Formula C7H6ClNO5S
Molecular Weight (g/mol) 251.64
MDL Number MFCD03094697
InChI Key QECYXMKYZQXEHM-UHFFFAOYSA-N
Synonym 2-methoxy-4-nitrobenzene-1-sulfonyl chloride, 2-methoxy-4-nitrobenzenesulfonylchloride, benzenesulfonyl chloride, 2-methoxy-4-nitro, pubchem11714, acmc-1ck5c, ksc207c7t, 2-chlorosulphonyl-5-nitroanisole, chloro 2-methoxy-4-nitrophenyl sulfone, 2-methoxy-4-nitrobenzensulfonyl chloride, 2-methoxy-4-nitrophenylsulfonyl chloride
PubChem CID 309458
SMILES COC1=CC(=CC=C1S(Cl)(=O)=O)[N+]([O-])=O

Specifications

Melting Point 96°C to 99°C
Quantity 1 g
UN Number UN3261
Beilstein 2136023
Sensitivity Moisture sensitive
IUPAC Name 2-methoxy-4-nitrobenzene-1-sulfonyl chloride
Formula Weight 251.65
Percent Purity 97%
Chemical Name or Material 2-Methoxy-4-nitrobenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.