missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-nitrophenyl isothiocyanate, 97%

Product Code. 11318645
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318645 5 g 5g
11308645 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318645 Supplier Thermo Scientific Alfa Aesar Supplier No. L12150.06
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 190774-55-1 | C8H6N2O3S | 210.207 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 190774-55-1
Molecular Formula C8H6N2O3S
Molecular Weight (g/mol) 210.207
MDL Number MFCD00041379
InChI Key NXWXXLFRMVILJN-UHFFFAOYSA-N
Synonym 2-methoxy-4-nitrophenyl isothiocyanate, 1-isothiocyanato-2-methoxy-4-nitro-benzene, acmc-20ans0, 2-methoxy4-nitrophenyl isothiocyanate, 2-methoxy-4-nitrophenylisothiocyanate, #, benzene, 1-isothiocyanato-2-methoxy-4-nitro, benzene, 1-isothiocyanato-2-methoxy-4-nitro-9ci
PubChem CID 610837
IUPAC Name 1-isothiocyanato-2-methoxy-4-nitrobenzene
SMILES COC1=C(C=CC(=C1)[N+](=O)[O-])N=C=S

Specifications

Melting Point 107°C to 109°C
Quantity 5 g
UN Number UN2923
Beilstein 6924925
Sensitivity Moisture sensitive
Formula Weight 210.21
Percent Purity 97%
Chemical Name or Material 2-Methoxy-4-nitrophenyl isothiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.