missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-vinylphenol, 97%, stab. with 0.01% BHT, Thermo Scientific™

Product Code. 11404614
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404614 5 g 5g
11494604 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11404614 Supplier Thermo Scientific Alfa Aesar Supplier No. A13194.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Methoxy-4-vinylphenol is used as a flavoring agent. It is also used as an aromatic substance resembling buckwheat. Further, it is used for chemical signaling in insects such as Red palm weevil. It is also utilized in imitation vanilla, coffee and cocoa.

Solubility
Miscible with water.

Notes
Air, light and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7786-61-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00015437
InChI Key YOMSJEATGXXYPX-UHFFFAOYSA-N
Synonym 2-methoxy-4-vinylphenol, 4-vinylguaiacol, 4-hydroxy-3-methoxystyrene, p-vinylguaiacol, phenol, 4-ethenyl-2-methoxy, 2-methoxy-4-vinyl-phenol, para-vinylguaiacol, guaiacol, 4-vinyl, phenol, 2-methoxy-4-vinyl, o-methoxy-p-vinylphenol
PubChem CID 332
ChEBI CHEBI:42438
IUPAC Name 4-ethenyl-2-methoxyphenol
SMILES COC1=C(C=CC(=C1)C=C)O

Specifications

Melting Point 25°C to 29°C
Density 1.11
Boiling Point 108°C to 110°C (3 mmHg)
Flash Point >100°C (212°F)
Odor Pleasant
Refractive Index 1.5818
Quantity 5 g
Beilstein 2044521
Solubility Information Miscible with water.
Formula Weight 150.18
Percent Purity 97%
Chemical Name or Material 2-Methoxy-4-vinylphenol, stabilized with 0.01% BHT
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.